Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VIP Mouse, Rat, DyLight 680, Clone: 2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105163FR
Description
VIP Monoclonal specifically detects VIP in Human, Rat, Porcine, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
VIP | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VIP | |
Vasoactive intestinal peptide adsorbed onto aluminum hydroxide gel | |
0.1 mL | |
Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
7432 | |
Human, Rat, Porcine, Bovine | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Binds to human, bovine, porcine or rat VIP, but does not bind to PHM or PACAP | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title