Learn More
VIPR1 Rabbit anti-Human, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 148781AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-172-9799
Add to cart
Description
Description
The Vasoactive Intestinal Polypeptide Receptor 1 (VPAC1) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.
Specifications
Specifications
VIPR1
Polyclonal
Unconjugated
VIPR1
HVR1, II, PACAP R 2, PACAP R2, PACAP type II receptor, RDC1, VAPC1, VIP R 1, VIPR, VIPR1, VIRG, VPAC1, VPCAP1R
Rabbit
Antigen Affinity Chromatography
RUO
7433
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P32241
VIPR1
VIPR1 Fusion Protein Ag6650
150 μL
Primary
Human
Antibody
IgG