Learn More
Cell Biolabs IncSupplier Diversity Partner VIRADUCTIN ADENO TRANSD KIT  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner AD200
ViraDuctin Adenovirus Transduction Reagent is designed to increase the transduction efficiency of adenoviral vectors in cells that lack the coxsackievirus-adenovirus receptor (CAR). 10 transductions/kit.
Catalog No. 50-672-461