Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Viral haemorrhagic septicemia virus ELISA Kit

Enzyme-linked immunosorbent assay for the detection of viral haemorrhagic septicemia virus (VHSV).

Manufacturer:  Abnova™ KA4894

Catalog No. 89-032-342


Add to cart

Description

Description

  • Used for the in vitro quantitative determination of human SOD1 in human serum, human plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant human IL-9
Specifications

Specifications

HRP
Qualitative
Viral haemorrhagic septicemia virus ELISA Kit
50 μL (Prediluted)
Colorimetric
Cell Culture Media, Homogenate
Viral haemorrhagic septicemia virus ELISA Kit is an enzyme-linked immunosorbent assay for the detection of viral haemorrhagic septicemia virus (VHSV).
Conjugate: 12mL
For research use only (RUO)
Store the kit at 4°C. Do not freeze.
1 Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit