Learn More
ZeptoMetrix Viral Lysate, Influenza A H1N1 2009 (Swine Flu)
Can be utilized as an antigen, as a source for the purification of viral proteins, or for the detection of viral antibodies
Specifications
Biosafety Level 2
Certifications/Compliance ISO, 13485
Detects Viral Antibodies
Format Solution
For Use With (Application) Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
22-157-672
View Documents
ZeptoMetrix
0810109N
Influenza A 2009 H1N1 (Swine Flu) Virus NY Strain Purified Viral Lysate This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
Description
Description
  • Propagated in the MDCK cell line
  • Purified using sucrose density gradient ultracentrifugation, disrupted in the presence of 0.5% Triton X-100 non-ionic detergent/0.6M KCl, and heat inactivated
Specifications
Specifications
2
Viral Antibodies
Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay
ISO, 13485
Solution
1 mg
Safety and Handling
Safety and Handling
ShelfLife : 1 Year