Learn More
Thermo Scientific? Virtuoso™ Vial Identification System
Eliminate errors from manual vial labeling. This advanced labeling-information device ensures accurate sample identification and increases productivity.
Manufacturer:  Thermo Scientific? 60180VT100
 View more versions of this product
Catalog No. 03-375-310
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
The Thermo Scientific? Virtuoso? Vial Identification System is an error-free option to manual vial labeling that turns a necessary chore into a value-added component within a laboratory workflow. With advanced labeling capabilities, the Virtuoso system ensures accurate vial identification and reliable data, and increases laboratory productivity. Use the system’s touch screen and network capabilities to transfer text, graphics, barcodes, and logos directly onto your vials.
Facilitates Your Labeling Requirements
  • Fully loaded with standard or fully user-defined templates
Increases Productivity
  • Eliminate error-prone manual vial labeling
  • Label a vial in just 5 seconds
  • Improve labeling efficiencies by downloading sample lists
  • Simplify vial sample identification
Produces High-Quality Labeling
  • Resists common laboratory solvents, blurring, and smudging
Provides an Advanced Labeling and information System in One Device
  • Designed by experienced analytical scientists
Description & Specifications
Specifications
Virtuoso™
Vial ID and Labelling
295mm
140mm
5A
Recommended Environment: +15?C to +30?C (+59?F to +86?F) Performance may deteriorate if operated outside recommended range. Recommended Storage: -25?C to +45?C (-13?F to 113?F) Short period only. Recommended Relative Humidity: Max. 80% RH up to 31°C Decreasing linearly to 50% RH at 40°C.
Pollution Level 2
300 dpi imaging system
3-5 seconds typical (full area print)
200mm
Universal power supply 100–240V AC
Virtuoso Vial Imaging System
24V DC, Over voltage category II
Promotions
Promotions
  • Thermo Scientific
    When you buy the Thermo Scientific™ Virtuoso™ Vial Identification System, you’ll get 10,000 Thermo Scientific™ SureStop™ Vials and Closures at no additional cost.
    09-30-2017