Learn More
Thermo Scientific™ Virtuoso™ Vial Identification System
Eliminate errors from manual vial labeling. This advanced labeling-information device ensures accurate sample identification and increases productivity.
Supplier: Thermo Scientific™ 60180VT407AP
Description
The Thermo Scientific™ Virtuoso™ Vial Identification System is an error-free option to manual vial labeling that turns a necessary chore into a value-added component within a laboratory workflow. With advanced labeling capabilities, the Virtuoso system ensures accurate vial identification and reliable data, and increases laboratory productivity. Use the system’s touch screen and network capabilities to transfer text, graphics, barcodes, and logos directly onto your vials.
Facilitates Your Labeling Requirements- Fully loaded with standard or fully user-defined templates
- Eliminate error-prone manual vial labeling
- Label a vial in just 5 seconds
- Improve labeling efficiencies by downloading sample lists
- Simplify vial sample identification
- Resists common laboratory solvents, blurring, and smudging
- Designed by experienced analytical scientists
Specifications
Virtuoso™ | |
Vial Identification System | |
Snap top amber vials, 11 mm blue snap caps and rubber/PTFE seals |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.