Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viruses and Bacteria Poster
Uses illustrations and actual photographs.
Manufacturer: Neo/Sci Corporation 351050
Description
- A close look at some of the most prevalent disease-causing viruses and bacteria
- Learn how to identify viruses and bacteria and how they harm humans Dimensions: 35W x 27 in.H (88.9 x 68.6cm)
Specifications
Science | |
27 in. | |
Closer look at the most prevalent disease causing viruses and bacteria known to man | |
Viruses and Bacteria Poster | |
88.9cm |
Illustrations and actual photographs | |
68.6cm | |
Paper | |
35 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title