Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Koehler™ Instrument Viscosity Reference Standards
SDP

For calibration and verification of kinematic and dynamic viscosity test equipment, both manual and automatic

Supplier:  Koehler™ Instrument 355004153

 View more versions of this product

Catalog No. 13-617-77


Add to cart

Description

Description

For calibration and verification of kinematic and dynamic viscosity test equipment, both manual and automatic

• Standards are based upon the NIST value of 1.0034 cSt for water at 20°C (68°F)
• Traceable to National Standards; manufactured according to ASTM D2162 specifications
• Supplied with ISO/IEC 17025 Certification Report
• Fully compliant with ASTM and related test procedures
• Calibrated to a precision of ±0.2% for viscosity and kinematic viscosity
• Each standard is calibrated at a minimum of five temperatures
• Supplied in 500mL amber bottle, with full certification and MSDS

Specifications

Specifications

Liquid
500 mL
ISO/IEC 17025, ASTM D2162, NIST
Kinematic and dynamic viscosity test equipment, both manual and automatic
13,994 cSt at 40°C
Viscosity Standard
For calibration and verification of kinematic and dynamic viscosity test
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit