Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High-Tech Conversions VISION 10™ Polyester Knit Cleanroom Wipers

High-Tech Conversions VISION 10™ Polyester Knit Cleanroom Wipers compatible with ISO 5 (Class 100) Controlled Environments

$745.00

Specifications

Autoclavable No
Color White
Disposable Yes
For Use With (Application) Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
Length (Metric) 23 cm
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
50-135-4699
View Documents
High-Tech Conversions
VS1098
9 x 8 in.
Case of 5000 for $745.00
Add to cart
 
50-135-4700
View Documents
High-Tech Conversions
VS1099/DEL
9 x 9 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Standard weight, knife-cut, polyester knit
  • Extremely low particle generation and chemical extractables
  • Double bagged and vacuum cleanroom packed
  • Meets USP 797 and USP 800 requirements
  • Lot to lot traceability
Specifications

Specifications

No
Yes
23 cm
Polyester Cleanroom Wipers
White
Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
Polyester
Non-sterile
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit