Learn More
High-Tech Conversions VISION 20™ Polyester Knit (Heavy-Weight) Cleanroom Wipers
High-Tech Conversions VISION 20™ Polyester Knit (Heavy-Weight) Cleanroom Wipers compatible with ISO 5 (Class 100) Controlled Environments
Manufacturer:  High-Tech Conversions VS201212/DEL
 View more versions of this product
Catalog No. 50-135-4702
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
This item is not returnable. View return policy
Description
Description
  • Heavy weight, knife-cut, polyester knit
  • Extremely low particle generation and chemical extractables
  • Double bagged
  • Vacuum cleanroom packed
  • Meets USP 797 and USP 800 requirements
  • Lot to lot traceability
Specifications
Specifications
30.5 cm
140 gsm
12 x 12 in.
30.5 cm
Cleanroom Wiper
Non-sterile
No
White
Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
Knit Polyester
Yes