Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High-Tech Conversions VISION 20™ Polyester Knit (Heavy-Weight) Cleanroom Wipers

High-Tech Conversions VISION 20™ Polyester Knit (Heavy-Weight) Cleanroom Wipers compatible with ISO 5 (Class 100) Controlled Environments

Specifications

Autoclavable No
Basis Weight 140 gsm
Color White
Disposable Yes
For Use With (Application) Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
50-110-7655
View Documents
High-Tech Conversions
VS2099/DEL
9 x 9 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
50-135-4702
View Documents
High-Tech Conversions
VS201212/DEL
12 x 12 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Heavy weight, knife-cut, polyester knit
  • Extremely low particle generation and chemical extractables
  • Double bagged
  • Vacuum cleanroom packed
  • Meets USP 797 and USP 800 requirements
  • Lot to lot traceability
Specifications

Specifications

No
White
Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
Knit Polyester
Non-sterile
140 gsm
Yes
23 cm
Cleanroom Wiper
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit