Thermo Scientific™ VISIONlite™ Spectrophotometer Software
Measure, analyze and export data quickly and easily using Thermo Scientific™ VISIONlite™ software.
Manufacturer: Thermo Scientific™ 869124400
 View more versions of this product
Catalog No. 14-380-604
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Applications Serves a variety of color determination needs, from industrial to pharmaceutical.
    Type Advanced Color Calculation Software
    Description Advanced Color Calculation Software
    Certifications/Compliance FDA guidelnes, IEEE
    For Use With Thermo Scientific UV-Visible spectrophotometers
    Includes User Manual
    Item Description Advanced Color Calculation Software
    Required Accessories System Requirements: MicrosoftWindows 2000/XP with Internet Explorer 5.0 or higher
    System Requirements Microsoft Windows 2000/XP with Internet Explorer 5.0 or higher
    Measure, analyze and export data quickly and easily using Thermo Scientific™ VISIONlite™ software. Designed for use with Thermo Scientific™ stand-alone visible and UV-visible spectrophotometers, user-friendly VISIONlite Software provides you with everything you need to acquire fast and accurate results.
    VISIONlite 5 software expands the capability of your instrument by offering method and data storage for these Thermo Scientific™ Spectrophotometers:
    SPECTRONIC™ 200 Spectrophotometer
    GENESYS™ 20 Visible Spectrophotometer
    GENESYS™ 10S UV-Visible and Visible Spectrophotometers
    Evolution 60S Spectrophotometer
    Evolution 300 Spectrophotometer
    VISIONlite 5 Software increases the capability of your instrument:
    • Add method and data storage for SPECTRONIC 200 Spectrophotometer and GENESYS 20 Visible Spectrophotometer.
    • Report results to 0.5nm wavelength resolution and 0.001A for SPECTRONIC 200 Spectrophotometer.
    • Perform automated kinetics measurements on SPECTRONIC 200 Spectrophotometer and GENESYS 20 Visible Spectrophotometer.
    • Scan on GENESYS 20 Visible Spectrophotometer.
    • Produce formatted reports in hard  copy or pdf format.
    VISIONlite ColorCalc Software turns your GENESYS 10S UV-Vis Spectrophotometer or Thermo Scientific™ Evolution™ 300 UV-Vis Spectrophotometer  into a powerful color analyser:
    • The Basic package offers all the most common colorspaces, illuminants and observer angles.
    • The Advanced package offers specialized calculations for yellowness, pharmaceuticals and quality control in wine production.
  • Description & Specifications
Related Products
Product Content Feedback