Learn More
Thermo Scientific™ VISIONlite™ Spectrophotometer Software
Measure, analyze and export data quickly and easily using Thermo Scientific™ VISIONlite™ software.
Manufacturer:  Thermo Scientific™ 869124300
 View more versions of this product
Catalog No. 14-380-605
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Measure, analyze and export data quickly and easily using Thermo Scientific™ VISIONlite™ software. Designed for use with Thermo Scientific™ stand-alone visible and UV-visible spectrophotometers, user-friendly VISIONlite Software provides you with everything you need to acquire fast and accurate results.
VISIONlite 5 software expands the capability of your instrument by offering method and data storage for these Thermo Scientific™ Spectrophotometers:
SPECTRONIC™ 200 Spectrophotometer
GENESYS™ 20 Visible Spectrophotometer
GENESYS™ 10S UV-Visible and Visible Spectrophotometers
Evolution 60S Spectrophotometer
Evolution 300 Spectrophotometer
VISIONlite 5 Software increases the capability of your instrument:
  • Add method and data storage for SPECTRONIC 200 Spectrophotometer and GENESYS 20 Visible Spectrophotometer.
  • Report results to 0.5nm wavelength resolution and 0.001A for SPECTRONIC 200 Spectrophotometer.
  • Perform automated kinetics measurements on SPECTRONIC 200 Spectrophotometer and GENESYS 20 Visible Spectrophotometer.
  • Scan on GENESYS 20 Visible Spectrophotometer.
  • Produce formatted reports in hard  copy or pdf format.
VISIONlite ColorCalc Software turns your GENESYS 10S UV-Vis Spectrophotometer or Thermo Scientific™ Evolution™ 300 UV-Vis Spectrophotometer  into a powerful color analyser:
  • The Basic package offers all the most common colorspaces, illuminants and observer angles.
  • The Advanced package offers specialized calculations for yellowness, pharmaceuticals and quality control in wine production.
Description & Specifications
Specifications
Serves a variety of color determination needs, from industrial to pharmaceutical.
Thermo Scientific UV-Visible spectrophotometers
Basic Color Calculation Software
Microsoft Windows 2000/XP with Internet Explorer 5.0 or higher
Basic Color Calculation Software
FDA guidelnes, IEEE
User Manual
System Requirements: MicrosoftWindows 2000/XP with Internet Explorer 5.0 or higher
Basic Color Calculation Software