Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Vismodegib Hedgehog inhibitor
May be dissolved in DMSO (200 mg/mL); or ethanol (10 mg/mL, warm)
Manufacturer: Novus Biologicals NBP26275825MG
Description
- Purity:>98% (HPLC); NMR (conforms)
- Molecular Weight: 421.3
- PubChem ID:24776445
Specifications
Powder | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Synthetic |
Off-white | |
25 mg | |
879085-55-9 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title