Learn More
VISTA/B7-H5/PD-1H Armenian Hamster anti-Mouse, DyLight 550, Clone: MH5A, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Supplier:  Novus Biologicals NBP311969R
Catalog No. NB151972
Add to Cart
Description
Description
VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Mouse samples. It is validated for ELISA, Immunohistochemistry
Specifications
Specifications
VISTA/B7-H5/PD-1H
Monoclonal
DyLight 550
50mM Sodium Borate
Armenian Hamster
Protein A purified
RUO
Primary
Mouse
IgG λ
ELISA, Immunohistochemistry
MH5A
ELISA, Immunohistochemistry
B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
The antibody was generated by immunising Armenian hamsters with mVISTA/B7-H5/PD-1HIg.
0.1 mL
Cancer, Cardiovascular Biology, Immunology
64115
Store at 4°C in the dark.