Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA Mouse anti-Human, PE, Clone: B7H5DS8, eBioscience™

Mouse Monoclonal Antibody

$140.00 - $357.00

Specifications

Antigen VISTA
Clone B7H5DS8
Concentration 5 μl/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12-108-841
Encompass
View Documents
eBioscience
12108841
25 Tests
Each for $140.00
Add to cart
 
12-108-842
Encompass
View Documents
eBioscience
12108842
100 Tests
Each for $357.00
Add to cart
 
Description

Description

Description: This B7H5DS8 monoclonal antibody recognizes human VISTA, also known as B7-H5 or PD-1H. This clone has been tested and shown not to cross-react with human B7-H4, B7-H6 and B7-H7. This B7H5DS8 antibody will work in flow cytometry only on live and not paraformaldehyde-fixed cells. Applications Reported: This B7H5DS8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B7H5DS8 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser

VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-H1, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans VISTA is expressed predomitly on hematopoietic cells with the highest densities on CD11b+ myeloid cells, and somewhat lower levels on CD4 and CD8 T cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. Mice deficient in VISTA are viable and have elevated frequencies of activated T cells but do not spontaneously develop autoimmune disorders.
Specifications

Specifications

VISTA
5 μl/Test
Monoclonal
Liquid
RUO
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
B7-H5, PD-H1, Gi24, Dies1, SISP1, C10orf54, PD-1H
VSIR
IgG1 κ
Affinity chromatography
Antibody
B7H5DS8
Flow Cytometry
PE
Mouse
Human
Q9H7M9
64115
VISTA-Fc protein
Primary
4° C, store in dark, DO NOT FREEZE!
VSIR
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit