Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA Monoclonal Antibody (MIH64), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Specifications

Antigen VISTA
Clone MIH64
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M035T02Y03
Encompass
View Documents
LIFE TECHNOLOGIES
M035T02Y03
25 μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
M035T03Y03
Encompass
View Documents
LIFE TECHNOLOGIES
M035T03Y03
100 μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

VISTA Monoclonal antibody specifically detects VISTA in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

VISTA
0.1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation
IgG2a κ
Affinity chromatography
Antibody
MIH64
Flow Cytometry
NovaFluor Yellow 610
Rat
Mouse
Q9D659
74048
Primary
4° C, store in dark, DO NOT FREEZE!
VSIR
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit