Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA Rat anti-Mouse, Brilliant Violet 421, Clone: MIH64, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 566272

 View more versions of this product

Catalog No. BDB566272


Add to cart

Description

Description

The MIH64 monoclonal antibody specifically binds to V-domain Ig suppressor of T cell activation (VISTA) which is also known as, Differentiation of ESCs 1 (Dies1). VISTA is a type I transmembrane glycoprotein that belongs to the Ig superfamily. VISTA has homology to the B7 family ligand, PD-L1 (CD274), and is likewise known as PD-1H. VISTA is expressed on naοve and activated T cells, natural killer (NK) cells, macrophages, dendritic cells, and neutrophils. VISTA functions as an inhibitory ligand. It can serve as a negative immune-checkpoint protein that suppresses T cell cytokine production and proliferation. It is also involved in regulating stem cell differentiation.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications

Specifications

VISTA
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Vista, PD-1H, Dies1
Mouse
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were
RUO
Primary
Murine
MIH64
Brilliant Violet 421
Affinity Purified
Rat
Rat IgG2a, κ
25μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit