Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA Rat anti-Mouse, PE, Clone: MIH64, BD

Rat Monoclonal Antibody

$129.00 - $293.00

Specifications

Antigen VISTA
Clone MIH64
Immunogen Mouse
Host Species Rat
Isotype Rat IgG2a, κ
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566270
View Documents
BD Biosciences
566270
25μg
Each for $129.00
Add to cart
 
BDB566269
View Documents
BD Biosciences
566269
100μg
Each for $293.00
Add to cart
 
Description

Description

The MIH64 monoclonal antibody specifically binds to V-domain Ig suppressor of T cell activation (VISTA) which is also known as, Differentiation of ESCs 1 (Dies1). VISTA is a type I transmembrane glycoprotein that belongs to the Ig superfamily. VISTA has homology to the B7 family ligand, PD-L1 (CD274), and is likewise known as PD-1H. VISTA is expressed on naοve and activated T cells, natural killer (NK) cells, macrophages, dendritic cells, and neutrophils. VISTA functions as an inhibitory ligand. It can serve as a negative immune-checkpoint protein that suppresses T cell cytokine production and proliferation. It is also involved in regulating stem cell differentiation.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
Specifications

Specifications

VISTA
Mouse
Rat IgG2a, κ
Affinity Purified
Monoclonal
Vista, PD-1H, Dies1
RUO
MIH64
Rat
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit