Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Pipette Tips: Low Retention Available on GSA/VA Contract for Federal Government customers only.

For use with Ovation and MLA Brand pipetters

$88.00 - $222.00

Specifications

Tip Style Low Retention
Compatibility Ovation™ M
Format Rack
For Use With (Application) Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications
View More Specs

Products 10
Catalog Number Mfr. No. Filtered Sterility Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Filtered Sterility Volume (Metric) Price Quantity & Availability  
22-146-563
View Documents
VistaLab™ Technologies
40701333LR
Filtered Sterile 2 µL
Case of 960 for $222.00
Add to cart
 
22-147-630
View Documents
VistaLab™ Technologies
40703004LR
Non-Filtered Non-sterile 1250 µL
Case of 960 for $133.00
Add to cart
 
22-146-560
View Documents
VistaLab™ Technologies
40701002LR
Non-Filtered Non-sterile 25 µL
Case of 960 for $88.00
Add to cart
 
22-147-631
View Documents
VistaLab™ Technologies
40703132LR
Non-Filtered Sterile 1250 µL
Case of 960 for $141.00
Add to cart
 
22-147-632
View Documents
VistaLab™ Technologies
40703332LR
Filtered Sterile 1250 µL
Case of 960 for $180.00
Add to cart
 
22-146-561
View Documents
VistaLab™ Technologies
40701032LR
Non-Filtered Sterile 25 µL
Case of 960 for $120.00
Add to cart
 
22-146-564 Available on GSA/VA Contract for Federal Government customers only.
View Documents
VistaLab™ Technologies
40702004LR
Non-Filtered Non-sterile 250 µL
Case of 960 for $88.00
Add to cart
 
22-146-565
View Documents
VistaLab™ Technologies
40702132LR
Non-Filtered Sterile 250 µL
Case of 960 for $123.00
Add to cart
 
22-146-562
View Documents
VistaLab™ Technologies
40701332LR
Filtered Sterile 25 µL
Case of 960 for $180.00
Add to cart
 
22-146-566 Available on GSA/VA Contract for Federal Government customers only.
View Documents
VistaLab™ Technologies
40702332LR
Filtered Sterile 250 µL
Case of 960 for $184.00
Add to cart
 
Description

Description

  • Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
  • Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications

Specifications

Low Retention
Ovation™ M
Rack
Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit