VistaLab Technologies™ MLA™ Brand D-Tipper™ Pipetters Available on GSA/VA Contract for Federal Government customers only.
Ideal for applications requiring dependable delivery of highly accurate volumes
Manufacturer: VistaLab Technologies™ 1146C
 View more versions of this product
Catalog No. 21-381-507C
  Check Availability
Add to Cart
    Specifications
    Accuracy ±2%
    Autoclavable Yes
    Certifications/Compliance ISO9001/2008
    Includes Glove guard
    Material Aluminum and Stainless Steel
    Warranty Lifetime
    Color Silver,Silver
    Model D-Tipper™
    Volume (Metric) 60μL
  • Description & Specifications
Related Products
Product Content Feedback