VistaLab Technologies™ MLA™ Brand Macro Pipetters Available on GSA/VA Contract for Federal Government customers only.

For large volume handling tasks such as reconstituting reagents, controls, or dilutions

Manufacturer: VistaLab Technologies™ 1077

 View more versions of this product

Catalog No. 22-146-313

  Check Availability
Add to Cart

    Specifications

    Accuracy ±0.6%
    Includes Graduated macro tip
    Material Aluminum and Stainless Steel
    Warranty Lifetime
    Color Red,Red
    Volume (Metric) 7mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit