VistaLab Technologies™ Ovation™ Electronic Single Channel Pipette Available on GSA/VA Contract for Federal Government customers only.

Ideal ergonomic choice for those needing a comprehensive menu of automated liquid handling functions

Manufacturer: VistaLab Technologies™ 10650125

 View more versions of this product

Catalog No. 22-146-101

  Check Availability
Add to Cart

    Specifications

    Accuracy ±0.8%
    Battery Type Lithium Ion
    Compatible Tips VistaLab™ tip
    Display LCD
    Finger Style Ergonomic
    For Use With (Equipment) VistaLab™ tip
    Increments 0.5μL
    Number of Speeds 5
    Warranty 1 Year
    Color Green,Green
    Model Right hand
    Number of Channels 1,1
    Volume (Metric) 2 to 125μL
    Speeds 5
    Compatibility VistaLab™ tip

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit