VistaLab Technologies™ Ovation™ Adjustable-Volume Pipetters Available on GSA/VA Contract for Federal Government customers only.

Naturally follows the contours of the palm

Manufacturer: VistaLab Technologies™ 10570200

 View more versions of this product

Catalog No. 22-146-604

  Check Availability
Add to Cart

    Specifications

    Accuracy ±0.8% at 200μL, ±0.8% at 100μL, ±2.5% at 20μL
    Display LCD
    For Use With (Application) Adjustable hook can be easily positioned for a custom fit for left- or right hand users
    Increments 0.2μL
    Color Blue,Blue
    Volume (Metric) 20 to 200μL
    Size Micro

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit