Learn More
VistaLab™ Technologies Ovation™ Adjustable-Volume Pipetters
SDP
Naturally follows the contours of the palm
Supplier:  VistaLab™ Technologies 10571000
 View more versions of this product
Catalog No. 22-146-605
Add to Cart
Description
Description
  • The adjustable hook can be easily positioned for a custom fit for left- or right-handed users
  • Fits comfortably in the palm of your hand
  • Volume setting is indicated by an LCD readout on the pipetter handle
  • Significantly improves postures and reduces forces associated with pipetting
  • Five popular volume settings can be programmed and recalled by pressing a single button
  • Energy release button stores the energy captured during the acquisition of tips and uses energy to push off the tip at the gentle push of a button
  • Low-force, form-fitting plunger allows user to manually control the speed of aspirating and dispensing with minimal effort
  • Nozzle with replaceable filter prevents contamination of the pipetter
  • Stands securely on the counter before, during, and after the job—no risk of contamination
  • In-lab calibration adjusted with internal software
Specifications
Specifications
±0.8% at 1000 μL, ±0.8% at 500 μL, ±3% (at 100 μL)
Left- or Right Hand Users
1
Adjustable Volume Pipette
LCD
CR2 battery, shelf hanger
1 Ea.
9 in.
3 in.
Purple
Polycarbonate ABS
Ovation™
100 to 1000 μL
Adjustable Hook Can be Easily Positioned for a Custom Fit for Left- or Right Hand Users
1 μL
2 Year Limited Warranty
7 in.
Replaceable CR2 Battery
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories