VistaLab Technologies™ Ovation™ Pipet Accessories: Reconditioning Nozzle Kit Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: VistaLab Technologies™ 90603011
 View more versions of this product
Catalog No. 22-146-253
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) 25 to 850μL, 25 to 1250μL Ovation™ BioNatural™ multichannel pipette
    Includes 2 Nozzles, 24 seals, 24 filters and tool
    Product Type Ovation™ reconditioning nozzle Kit
  • Description & Specifications
Related Products
Product Content Feedback