Learn More
VistaLab Technologies™ Pipet Volume Calibration Kits Available on GSA/VA Contract for Federal Government customers only.
For discrete-sample and flow-through spectrophotometers
Manufacturer:  VistaLab Technologies™ 9082
 View more versions of this product
Catalog No. 21-381-3
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Calibration kit
To test the delivered volume of any laboratory pipette or diluter within a range of 10 to 1000μL
Sealed cuvets of four standards for establishing curves, vials of calibration reagent of known concentration, covering testing range of 10 to 1000μL, calibration cuvets containing diluents, straight edge ruler, quality control record sheets
For discrete sample spectrophotometers, mechanical pipetters/Glass pipets
10 to 1000μL
100 Tests