Learn More
VistaLab Technologies™ Pipet Volume Calibration Kits
For discrete-sample and flow-through spectrophotometers
Manufacturer:  VistaLab Technologies™ 9083
Catalog No. 21-381-4
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Includes:
  • Sealed cuvets of four standards for establishing curves
  • Vials of calibration reagent of known concentration, covering testing range of 10 to 1000μL
  • Calibration cuvets containing diluent
  • Straight-edge ruler
  • Quality control record sheet
  • Graph paper
Description
Description
For mechanical pipetters or glass pipets, volume range 10 to 1000μL.
  • No special scales are needed
Order Info
Discrete-sample kits are for use with models accepting 12mm diameter sample cuvets.
Specifications
Specifications
Calibration Kit
To Test The Delivered Volume of Any Laboratory Pipette or Diluter Within a Range of 10 to 1000 µL
Sealed cuvets of four standards for establishing curves, vials of calibration reagent of known concentration, covering testing range of 10 to 1000 µL, calibration cuvets containing diluents, straight edge ruler, quality control record sheets
For Flow Through Spectrophotometers
10 to 1000 µL
100 Tests