VistaLab Technologies™ Pipet Volume Calibration Kits Available on GSA/VA Contract for Federal Government customers only.
For discrete-sample and flow-through spectrophotometers
Manufacturer: VistaLab Technologies™ 9081
 View more versions of this product
Catalog No. 21-381-2
  Check Availability
Add to Cart
    Specifications
    Product Type Calibration kit,Calibration kit
    For Use With (Equipment) For flow through spectrophotometers
    For Use With (Application) To test the delivered volume of any laboratory pipette or diluter within a range of 10 to 1000μL
    Volume (Metric) 10 to 1000μL
    Includes Sealed cuvets of four standards for establishing curves, vials of calibration reagent of known concentration, covering testing range of 10 to 1000μL, calibration cuvets containing diluents, straight edge ruler, quality control record sheets
    Quantity 30 Tests
  • Description & Specifications
Related Products
Product Content Feedback