Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Pipette Tips: Sterile, Filtered
GREENER_CHOICESDP

For use with Ovation and MLA Brand pipetters

$185.00 - $246.00

Specifications

Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Blue
Autoclavable Not Autoclavable
View More Specs

Products 4
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
22-146-359
GREENER_CHOICESDP
View Documents
VistaLab™ Technologies
40601332
25 μL
Case of 960 for $185.00
Edge
Add to Cart
 
22-146-360
GREENER_CHOICESDP
View Documents
VistaLab™ Technologies
40601333
2 μL
Case of 960 for $246.00
Edge
Add to Cart
 
22-146-361
GREENER_CHOICESDP
View Documents
VistaLab™ Technologies
40602332
250 μL
Case of 960 for $208.00
Add to Cart
 
22-146-362
GREENER_CHOICESDP
View Documents
VistaLab™ Technologies
40603332
1250 μL
Case of 960 for $246.00
Add to Cart
 
Description

Description

  • Sterile tips packaged in VistaRaks or Individually Wrapped
  • VistaRak: 5 racks, 192 tips/rack
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time

Order Info

Packaged 192 tips/rack, 5 racks/case

Specifications

Specifications

Filtered
Clear
Not Autoclavable
DNase, RNase, and Pyrogen free
Rack
VistaRak: 5 racks
Single-use
Polypropylene, Polyethylene (Filter)
Sterile
Blue
5 in.
e-beam
Straight
192 Tips/Rack
No
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.