Learn More
Vistek™ Vibration Isolation Platforms
Significantly improve the performance of microscopes, lab balances, and other sensitive instruments
Manufacturer:  NEWPORT CORPORATION VIP32012181555
 View more versions of this product
Catalog No. 10-999-604
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
For general purpose, precision applications, improve the performance of microscopes, lab balances, and other sensitive instruments
15 to 55 lb.
12 x 18 in.
6.8Hz
Vibration Isolation Platform
Inverted or other types of microscopes, inspection tools
6.8 to 24.9kg
30.48 x 45.72cm
Steel