Learn More
Novus Biologicals™ VisUCyte™ Antigen Retrieval Reagent-Universal
VisUCyte™ Antigen Retrieval Reagents are three different antigen retrieval systems that utilize heat-induced recovery of cell and tissue antigens.
Manufacturer:  Novus Biologicals™ NBVCTS023
Catalog No. NBVCTS023
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • The optimal antigen unmasking condition is a function of temperature (90-100°C) and incubation time (up to 30 minutes) and should be established by each individual investigator.
Specifications
Specifications
50 mL
Store at 4°C.
VisUCyte(TM) Antigen Retrieval Reagent-Universal