Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Vitamin A acetate in gelatin, 500,000 I.U./g

Click to view available options
Quantity:
25g
100g
500g
127-47-9
C22H32O2
328.496
MFCD00019413
QGNJRVVDBSJHIZ-QHLGVNSISA-N
retinyl acetate,vitamin a acetate,retinol acetate,retinol, acetate,all-trans-retinyl acetate,crystalets,vitamin a1 acetate,all-trans-retinol acetate,vitamin a alcohol acetate,davitan a 650
638034
CHEBI:32095
[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C
127-47-9
C22H32O2
328.496
MFCD00019413
QGNJRVVDBSJHIZ-QHLGVNSISA-N
retinyl acetate,vitamin a acetate,retinol acetate,retinol, acetate,all-trans-retinyl acetate,crystalets,vitamin a1 acetate,all-trans-retinol acetate,vitamin a alcohol acetate,davitan a 650
638034
CHEBI:32095
[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C
Sensitivity Air and light sensitive; Hygroscopic
Quantity 500g
Beilstein 1915439
Merck Index 14,10013
Concentration or Composition (by Analyte or Components) 500,000 IU/g
Chemical Name or Material Vitamin A acetate, in gelatin

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360D

EINECSNumber : 204-844-2

RTECSNumber : VH6825000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen