Learn More
MP Biomedicals Vitamin B12 Deficient Diet, Modified, Powder
Vitamin B12 deficient diet. Contains vitamins and minerals.
Manufacturer:  MP Biomedicals 0296017210
 View more versions of this product
Catalog No. MP296017210
Add to cart
Description
Description
  • Diet consisting of casein, iodinated casein, choline, salt mixture no
  • 2 USP and vitamin mix
  • Ideal for experiments containing control vs. modified diets
Specifications
Specifications
0.3048 m
Autoclavable
Powder
Non-sterile
Animal Diet
1 ft.
Yes
10 kg
Room Temperature
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.