Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin D BP Mouse anti-Human, Alexa Fluor™ 405, Clone: 4B9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223535AF405

Catalog No. NP2233A40


Add to cart

Description

Description

Vitamin D BP Monoclonal antibody specifically detects Vitamin D BP in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Vitamin D BP
4B9
Western Blot, ELISA
DBP/GC, DBPVDBG, EC 6.3.1.5, Gc-globulin, GRD3, Group-specific component, group-specific component (vitamin D binding protein), hDBP, VDB, VDBP, vitamin D-binding alpha-globulin, vitamin D-binding protein
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone 4B9 is specific for human Gc-globulin
ELISA, Western Blot
Alexa Fluor™ 405
50 mM sodium borate with 0.05% sodium azide
GC
Gc-globulin isolated from human plasma adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
2638
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit