Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin D BP Mouse anti-Human, Alexa Fluor™ 594, Clone: 2G2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223541AF594

Catalog No. NP22341A94

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Vitamin D BP Monoclonal antibody specifically detects Vitamin D BP in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Vitamin D BP
2G2
Western Blot, ELISA
DBP/GC, DBPVDBG, EC 6.3.1.5, Gc-globulin, GRD3, Group-specific component, group-specific component (vitamin D binding protein), hDBP, VDB, VDBP, vitamin D-binding alpha-globulin, vitamin D-binding protein
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone 2G2 is specific for human Gc-globulin.
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
GC
Gc-globulin isolated from human plasma adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
2638
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit