Learn More
Vitamin D Receptor (VDR) Ab-1 Rat Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Vitamin D Receptor (VDR) Ab-1, Rat Monoclonal Antibody.
Supplier: Epredia RT200P1
Description
VDR is a member of super family of steroid hormone receptors. It is known to play role in cellular growth and differentiation, calcium and phosphorus metabolism and bone remodeling. Human breast carcinomas, leukemias, and prostatic carcinoma cells express VDR, which can modify their growth and differentiation. In avians, VDR appears as a doublet of 58kDa and 60kDa.Host Species: Rat
Clone: 9A7γ.E10.E4
Isotype: IgG2b
Species Reactivity: Human, Pig, Mouse, Rat, Hamster, Frog, Chicken and Fish. Others not known.
Epitope: aa 89-105 (near the DNA-binding domain)
Immunogen: Vitamin D receptor protein from chicken intestine
Molecular Weight: 53kDa
Positive Control: LNCAP cells, HeLa cells
Cellular Localization: Nuclear Ab-1 reacts with both the occupied and unoccupied forms of VDR and shows no cross reaction with estrogen and glucocorticoid receptors. It is excellent for biochemical and histochemical applications.
Recommended for:
- Western Blotting
- Gel Supershift Assay
Note:
USA: RUO; Int'l: RUO
Specifications
Vitamin D Receptor (VDR) Ab-1 | |
Monoclonal | |
200µg/mL | |
Rat | |
500 μL | |
Nuclear Receptors | |
Chicken, Fish, Frog, Hamster, Human, Mouse, Porcine | |
IgG2b |
Western Blot | |
9A7gamma.E10.E4 | |
Unconjugated | |
Vitamin D receptor protein from chicken intestine | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.