Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin D3, ≥99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
67-97-0
C27H44O
384.648
MFCD00078131
QYSXJUFSXHHAJI-YRZJJWOYSA-N
vitamin d3,cholecalciferol,calciol,colecalciferol,oleovitamin d3,arachitol,activated 7-dehydrocholesterol,vitamin d,colecalcipherol,colecalciferolum
5280795
CHEBI:28940
(1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C
67-97-0
C27H44O
384.648
MFCD00078131
QYSXJUFSXHHAJI-YRZJJWOYSA-N
vitamin d3,cholecalciferol,calciol,colecalciferol,oleovitamin d3,arachitol,activated 7-dehydrocholesterol,vitamin d,colecalcipherol,colecalciferolum
5280795
CHEBI:28940
(1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C
Specific Rotation +105° to +112° (25°C, 589nm) (c=0.8, ethanol) (1 dm, within 30 min.)
Specific Rotation Condition +108.50 (25.00°C c=0.8,ethanol)
Quantity 1g
Color White
Packaging Glass bottle
Melting Point 82°C to 87°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Merck Index 15, 10216
Formula Weight 384.64
Physical Form Crystalline Powder or Crystals
Percent Purity ≥98.5%
Chemical Name or Material Vitamin D3
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Causes damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger