Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin K1, 97.0 to 103.0%, Affymetrix/USB™

Click to view available options
Quantity:
5g
10g
Packaging:
Plastic bottle
84-80-0
C31H46O2
450.707
MFCD00214063
MBWXNTAXLNYFJB-NKFFZRIASA-N
vitamin k1,phytonadione,phylloquinone,phytylmenadione,phyllochinon,phytomenadione,alpha-phylloquinone,3-phytylmenadione,konakion,2-methyl-3-phytyl-1,4-naphthoquinone
5284607
CHEBI:18067
2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C
84-80-0
C31H46O2
450.707
MFCD00214063
MBWXNTAXLNYFJB-NKFFZRIASA-N
vitamin k1,phytonadione,phylloquinone,phytylmenadione,phyllochinon,phytomenadione,alpha-phylloquinone,3-phytylmenadione,konakion,2-methyl-3-phytyl-1,4-naphthoquinone
5284607
CHEBI:18067
2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C
Specific Gravity 0.984
Packaging Plastic bottle
Flash Point >110°C (230°F)
Quantity 10g
Boiling Point ~140°C (0.001mm)
Melting Point ~-20°C
Formula Weight 450.70
Physical Form Viscous Liquid
Percent Purity 97.0 to 103.0%
Chemical Name or Material Vitamin K1