Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Vitamin K3 Monoclonal Antibody (5H9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549644
Description
Antibody tested in indirect ELISA.
Vitamin K3, also known as menadione, is a synthetic form of vitamin K that is used as a nutritional supplement. It is commonly used in animal feed to prevent vitamin K deficiency and improve overall health and growth. Vitamin K3 is also used in the production of various food and pharmaceutical products. It plays a critical role in blood clotting and bone metabolism, and it has been shown to have antioxidant and anti-inflammatory properties. However, excessive intake of vitamin K3 can be toxic and can cause liver damage.
Specifications
Vitamin K3 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Mouse | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Lateral Flow | |
5H9 | |
Unconjugated | |
menadione | |
Conjugate of VK3 hapten and KLH. | |
200 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction