Thermo Scientific™ Vitek™ QC Sets

Obtain faster recovery, enhanced viability, and direct-streak-to-plate capabilities with QC organisms manufactured using an unmatched proprietary gel preservation process.

Manufacturer: Thermo Scientific™ R4682010

 View more versions of this product

Catalog No. 22-246-502

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    ATCC® 19414™*,19615™*,29212™*,29971™*,6056™*,9528™*,9809™*
    Type Enterococcus durans,Enterococcus durans,Enterococcus faecalis,Erysipelothrix rhusiopathiae,Staphylococcus xylosus,Streptococcus equi subsp. equi,Streptococcus gallolyticus,Streptococcus pyogenes
    Test Type Qualitative QC
    Target Organism Class Organism Sets
    Certifications/Compliance ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
    Quantity 5 Loops/Pk.,5 Loops/Pk.
    Storage Requirements 2-8°C
    Product Type Streptococcus gallolyticus
    Description Vitek™ GPI QC 1 Loop Set/7
    Format Culti-Loops™
    Storage 2-8°C

    Ensure high-performance products, reproducible clinical test results and consistent microbiological evaluation of pharmaceuticals and food with Thermo Scientific™ Culti-Loops™ quality control organisms. Thermo Scientific™ Vitek™ QC Sets include a ready-to-use set of QC organisms in Culti-Loops format which are ready-to-use, disposable bacteriological loops containing stabilized, viable microorganisms in a unique gel matrix.

    Trust Thermo Scientific quality control organisms for quality results you can depend on, backed by over 30 years of expertise in the manufacturing and packaging of controlled microorganisms.

    Convenience

    • Decrease time and unnecessary costs of laboratory resources with a direct-streak inoculating loop that requires no rehydration.
    Performance
    • Obtain enhanced viability and recovery with proprietary gel-preserved organisms.
    Value
    • Consolidate ordering time and shipping costs by turning to one supplier for high-quality culture media and quality control organisms.
    Confidence
    • Reach complete traceability with ATCC® License Derivative strains.
    Efficiency
    • Achieve accurate and convenient documentation with peel-off, transferable labels.
    Safety
    • Reduce the risk of contamination and infection with zero organism handling.
    • More than 600 quality control organisms, including convenient sets for commonly used identification systems
    • ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
    • ATCC® Licensed Derivative Program member
    Not all products are available for sale in all territories. Please inquire.

    Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

    *The ATCC Licensed Derivative® Emblem, the ATCC Licensed Derivative® word mark, and the ATCC catalog marks are trademarks of ATCC®. Thermo Fisher Scientific is licensed to use these trademarks and to sell products derived from ATCC® cultures.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit