Learn More
Vitronectin Mouse anti-Bovine, Janelia Fluor 646, Clone: A18, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223571JF646
Catalog No. NBP23571646
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Vitronectin Monoclonal antibody specifically detects Vitronectin in Bovine samples. It is validated for Immunohistochemistry.
Specifications
Specifications
Vitronectin
A18
Immunohistochemistry
Purified
VTN
Lysed bovine corneal endothelial cells and extracellular matrix
Protein A or G purified
Immunology, Innate Immunity
Primary
7448
Bovine, Equine, Rabbit
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Clone A18 is highly specific for vitronectin. There is no evidence for cross-reactivity with other connective tissue proteins (fibronectin, elastin, collagen, laminin). Clone A18 cross-reacts with rabbit and horse vitronectin, no reactivity with human or sheep.