Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitronectin Mouse anti-Human, Bovine, Canine, Equine, Feline, Goat, Alexa Fluor™ 750, Clone: HV23, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223526AF750

Catalog No. NP22326A70


Add to cart

Description

Description

Vitronectin Monoclonal antibody specifically detects Vitronectin in Human, Bovine, Canine, Equine, Feline, Goat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Vitronectin
HV23
Western Blot, ELISA
Conjugated, Purified
VTN
Human vitronectin purified from plasma by heparin-affinity chromatography
Protein A or G purified
Immunology, Innate Immunity
Primary
7448.0
Bovine, Canine, Equine, Feline, Goat, Human
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Clone HV23 is highly specific for vitronectin. There is no evidence for cross-reactivity with other connective tissue proteins (fibronectin, elastin, collagen, laminin). Clone HV23 cross-reacts with vitronectin from cat, dog, goat, cow and to a lesser extent with horse.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit