Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitronectin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 720229

Encompass

Catalog No. 72-022-9


Add to cart

Description

Description

This antibody is predicted to react with Monkey, Dog, Mouse

Vitronectin Polyclonal antibody specifically detects Vitronectin in Human samples. It is validated for Western Blot
Specifications

Specifications

Vitronectin
Western Blot
0.5 mg/mL
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4
P04004
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7448
Liquid
Polyclonal
Unconjugated
VTN
Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN
Peptides corresponding to human VTN (aa367-384, 107-120)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit