Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Vitronectin / VTN Protein, Tag Free, ultra sensitivity (primary amine labeling)
Supplier: ACROBiosystems VINH811325UG
Recombinant Protein;25UG;Biotinylated Human Vitronectin, Tag Free, primary amine labeling (VIN-H8113) is expressed from E.coli cells. It contains AA Asp 20 - Arg 398 (Accession # P04004-1).;This protein carries no "tag". The protein has a calculated MW of 43.2 kDa. The protein migrates as 47-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Vitronectin, Tag Free, primary amine labeling (Cat. No. VIN-H8113) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human ITGAV&ITGB5 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT5-H52W5) with a linear range of 0.156-1.25 μg/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.