Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Vivaspin™ 20 Sample Concentrators: PROMO

Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. GE Healthcare Life Vivaspin™ 20 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.

Manufacturer:  GE Healthcare 28932361PM

 View more versions of this product

Catalog No. 45-001-580LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.

  • Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
  • Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
  • Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
  • Compatible pH range: 1 to 9
  • Active membrane area: 6.0cm2

For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.

Specifications

Specifications

Polyethersulfone
30,000 d
6cm2
30mm
116mm
25/Pk.
5 to 20mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit