Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivaspin Turbo 15RC

Catalog No. 01288115 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
5 kDa
50 kDa
Quantity:
12 Pk.
48 Pk.
Catalog No. MWCO Quantity
01-288-115 5 kDa 48 Pk.
01-288-118 10 kDa 12 Pk.
01-288-117 10 kDa 48 Pk.
01-288-116 5 kDa 12 Pk.
01-288-113 50 kDa 12 Pk.
01-288-112 50 kDa 48 Pk.
01-288-110 100 kDa 48 Pk.
7 options

Catalog No. 01-288-115

Supplier: Sartorius VS15TR12

Only null left
Add to Cart
Add to Cart

The highly optimized design of Vivaspin Turbo 15RC with twin vertical membranes brings ultrafast speeds to your ultrafiltration processes. The low adsorbing regenerated cellulose (RC) membranes and smooth transition between filter and housing ensure maximum target molecule recoveries every time.

  • Fastest sample concentration with maximum target molecule recoveries
  • Simplest, pipette-friendly retentate retrieval
  • Highest chemical and temperature resistance
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Format Spin Column
For Use With (Equipment) Centrifuge
Material SBC
Material (Membrane) Regenerated Cellulose
MWCO 5 kDa
Sample Volume 4 to 15 mL
Sterility Non-sterile
Quantity 48 Pk.
Type Centrifugal
Volume (Metric) 15 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.