Learn More
Athens Research And Technology IncSupplier Diversity Partner VLDL LIPOPROTEINS HU 5MG DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 12162212045MG
Lipoproteins, Very Low Density, Human Plasma: 5x1 MG; Liquid in 150 mM NaCl, pH 7.4, and 0.1% EDTA; MW 10-80,000,000. Native; Prepared and purified from fresh, non-frozen human plasma in the Athens Research & Technology laboratory. Lipoproteins, Molecular Biology Reagent
Catalog No. 50-489-872