Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VLDLR Mouse anti-Human, Rat, PE/Cy5.5, Clone: VLDLR/1337, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254553PECY55

Catalog No. N25453PECY5


Add to cart

Description

Description

VLDLR Monoclonal antibody specifically detects VLDLR in Human, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

VLDLR
VLDLR/1337
Flow Cytometry
Conjugated, Purified
VLDLR
Recombinant human VLDLR fragment from c-terminal (exact sequence is proprietary)
Protein A or G purified
Apoptosis, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary
7436.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit